4-Bromo-3,5-dichlorofluorobenzene 98%

Synonyme: 2-Bromo-1,3-dichloro-5-fluorobenzene

BD161309-1G BD161309-5G BD161309-25G BD161309-100G BD161309-250MG
BLDPBD161309-1GEA 19 EUR
BLDPBD161309-1G BLDPBD161309-5G BLDPBD161309-25G BLDPBD161309-100G BLDPBD161309-250MG
4-Bromo-3,5-dichlorofluorobenzene 98%
4-Bromo-3,5-dichlorofluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₂BrCl₂F
Molecular Weight: 243,9 g/mol
Lagertemperatur: Ambient
MDL: MFCD11845890
CAS-Nummer: 263333-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR