2,6-Dibromonitrobenzene 97%

Synonyme: 1,3-Dibromo-2-nitrobenzene

BD161304-10G BD161304-5G BD161304-25G BD161304-100G BD161304-250MG BD161304-1G
BLDPBD161304-10GEA 162 EUR
BLDPBD161304-10G BLDPBD161304-5G BLDPBD161304-25G BLDPBD161304-100G BLDPBD161304-250MG BLDPBD161304-1G
2,6-Dibromonitrobenzene 97%
2,6-Dibromonitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₃Br₂NO₂
Molecular Weight: 280,902 g/mol
Lagertemperatur: Ambient
MDL: MFCD09998726
CAS-Nummer: 13402-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR