Methyl 4-amino-2,5-dimethylbenzoate 95%

BD161288-250MG
BLDPBD161288-250MGEA 30 EUR
BLDPBD161288-250MG
Methyl 4-amino-2,5-dimethylbenzoate 95%
Methyl 4-amino-2,5-dimethylbenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Ambient
MDL: MFCD09839220
CAS-Nummer: 21339-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR