3-Chlor-4-fluorzimtsäure 97%

Synonyme: 3-(3-Chlor-4-fluorphenyl)propensäure3-Chlor-4-fluorcinnamsäure

BLDPBD161277-10GEA 98 EUR
BLDPBD161277-10G BLDPBD161277-25G BLDPBD161277-1G BLDPBD161277-5G
3-Chlor-4-fluorzimtsäure 97%
3-Chlor-4-fluorzimtsäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₆ClFO₂
Molecular Weight: 200,6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01631393
CAS-Nummer: 155814-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR