3,5-Dichloro-4-fluoroaniline 98%

BD161261-5G BD161261-1G BD161261-100G BD161261-10G BD161261-250MG BD161261-25G
BLDPBD161261-5GEA 16 EUR
BLDPBD161261-5G BLDPBD161261-1G BLDPBD161261-100G BLDPBD161261-10G BLDPBD161261-250MG BLDPBD161261-25G
3,5-Dichloro-4-fluoroaniline 98%
3,5-Dichloro-4-fluoroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄Cl₂FN
Molecular Weight: 180,007 g/mol
Lagertemperatur: Ambient
MDL: MFCD09835492
CAS-Nummer: 2729-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR