5-Brom-2-(trifluormethoxy)anilin 98%

Synonyme: 3-Amino-4-(trifluoromethoxy)bromobenzene

BD161256-100G BD161256-10G BD161256-25G BD161256-500G BD161256-5G
BLDPBD161256-100GEA 105 EUR
BLDPBD161256-100G BLDPBD161256-10G BLDPBD161256-25G BLDPBD161256-500G BLDPBD161256-5G
5-Brom-2-(trifluormethoxy)anilin 98%
5-Brom-2-(trifluormethoxy)anilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅BrF₃NO
Molecular Weight: 256,0200 g/mol
Lagertemperatur: Ambient
MDL: MFCD04039242
CAS-Nummer: 886762-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR