(2-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%

Synonyme: 2-Fluoro-4-(trifluoromethyl)phenylacetic acid

BD161249-100MG BD161249-250MG BD161249-1G
BLDPBD161249-100MGEA 30 EUR
BLDPBD161249-100MG BLDPBD161249-250MG BLDPBD161249-1G
(2-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%
(2-Fluor-4-(trifluormethyl)phenyl)essigsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
Lagertemperatur: Ambient
MDL: MFCD00061186
CAS-Nummer: 209991-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR