2-(4-Fluoro-3-nitrophenyl)acetic acid 98%

BD161246-100MG BD161246-250MG BD161246-10G BD161246-25G BD161246-100G BD161246-5G BD161246-1G
BLDPBD161246-100MGEA 9 EUR
BLDPBD161246-100MG BLDPBD161246-250MG BLDPBD161246-10G BLDPBD161246-25G BLDPBD161246-100G BLDPBD161246-5G BLDPBD161246-1G
2-(4-Fluoro-3-nitrophenyl)acetic acid 98%
2-(4-Fluoro-3-nitrophenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆FNO₄
Molecular Weight: 199,1400 g/mol
Lagertemperatur: Ambient
MDL: MFCD09743463
CAS-Nummer: 192508-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR