2-Fluoro-5-methoxyaniline 97%

Synonyme: 3-Amino-4-fluoroanisole,6-Fluoro-m-anisidine

BD161236-1G BD161236-5G BD161236-10G BD161236-25G BD161236-100G
BLDPBD161236-1GEA 37 EUR
BLDPBD161236-1G BLDPBD161236-5G BLDPBD161236-10G BLDPBD161236-25G BLDPBD161236-100G
2-Fluoro-5-methoxyaniline 97%
2-Fluoro-5-methoxyaniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₈FNO
Molecular Weight: 141,143 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11226283
CAS-Nummer: 62257-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR