(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 97%

Synonyme: Boc-Tyr(Me)-OH,N-Boc-O-methyl-L-tyrosine

BD16123-100G BD16123-5G BD16123-25G BD16123-1G
BLDPBD16123-100GEA 477 EUR
BLDPBD16123-100G BLDPBD16123-5G BLDPBD16123-25G BLDPBD16123-1G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,3 g/mol
Lagertemperatur: Ambient
MDL: MFCD00065603
CAS-Nummer: 53267-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR