2-Methoxy-5-(trifluormethoxy)benzylalkohol 95%

BD161226-250MG BD161226-1G BD161226-5G
BLDPBD161226-250MGEA 24 EUR
BLDPBD161226-250MG BLDPBD161226-1G BLDPBD161226-5G
2-Methoxy-5-(trifluormethoxy)benzylalkohol 95%
2-Methoxy-5-(trifluormethoxy)benzylalkohol

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉F₃O₃
Molecular Weight: 222,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD04115960
CAS-Nummer: 562840-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR