4-Bromo-2-chloro-3-fluorophenol 95%

BD161218-100MG BD161218-250MG BD161218-1G
BLDPBD161218-100MGEA 32 EUR
BLDPBD161218-100MG BLDPBD161218-250MG BLDPBD161218-1G
4-Bromo-2-chloro-3-fluorophenol 95%
4-Bromo-2-chloro-3-fluorophenol

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₃BrClFO
Molecular Weight: 225,4428 g/mol
Lagertemperatur: Ambient
MDL: MFCD11849932
CAS-Nummer: 1233026-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR