2-chloro-1,3-dimethoxybenzene 98%

BLDPBD161217-100MGEA 8 EUR
BLDPBD161217-100MG BLDPBD161217-250MG
2-chloro-1,3-dimethoxybenzene 98%
2-chloro-1,3-dimethoxybenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₉ClO₂
Molecular Weight: 172,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835196
CAS-Nummer: 7051-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR