2-Brom-4-iodanisol 95%
|
|
BLDPBD161207-100MGEA
15
EUR
BLDPBD161207-100MG
BLDPBD161207-1G
BLDPBD161207-5G
BLDPBD161207-10G
BLDPBD161207-250MG
2-Brom-4-iodanisol 95%
2-Brom-4-iodanisol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₆BrIO Molecular Weight: 312,9304 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425935 CAS-Nummer: 182056-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...