2-Chloro-5-hydroxybenzonitrile 97%

BLDPBD161164-1GEA 135 EUR
BLDPBD161164-1G BLDPBD161164-250MG BLDPBD161164-100MG
2-Chloro-5-hydroxybenzonitrile 97%
2-Chloro-5-hydroxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄ClNO
Molecular Weight: 153,566 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743462
CAS-Nummer: 188774-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR