Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate 95%

Synonyme: Ethyl 2,6-dichloro-5-nitropyrimidine-4-carboxylate

BD161103-100MG BD161103-5G BD161103-250MG BD161103-1G
BLDPBD161103-100MGEA 31 EUR
BLDPBD161103-100MG BLDPBD161103-5G BLDPBD161103-250MG BLDPBD161103-1G
Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate 95%
Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₅Cl₂N₃O₄
Molecular Weight: 266,04 g/mol
Lagertemperatur: Freezer
MDL: MFCD00831638
CAS-Nummer: 54368-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR