N-Boc-3-methyl-L-phenylalanine 98%

Synonyme: Boc-Phe(3-Me)-OH

BD16105-250MG BD16105-1G
BLDPBD16105-250MGEA 21 EUR
BLDPBD16105-250MG BLDPBD16105-1G
N-Boc-3-methyl-L-phenylalanine 98%
N-Boc-3-methyl-L-phenylalanine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,336 g/mol
Lagertemperatur: Ambient
MDL: MFCD01862944
CAS-Nummer: 114873-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR