Boc-Phe-OH 98%

BLDPBD16099-5GEA 7 EUR
BLDPBD16099-5G BLDPBD16099-100G BLDPBD16099-25G BLDPBD16099-500G BLDPBD16099-10G BLDPBD16099-0.5G
Boc-Phe-OH 98%
Boc-Phe-OH

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₉NO₄
Molecular Weight: 265,309 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00558969
CAS-Nummer: 4530-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR