(R)-2-(Bromomethyl)-2,3-dihydrobenzo[b][1,4]dioxine 95+%

BD160937-1G BD160937-100MG BD160937-250MG
BLDPBD160937-1GEA 221 EUR
BLDPBD160937-1G BLDPBD160937-100MG BLDPBD160937-250MG
(R)-2-(Bromomethyl)-2,3-dihydrobenzo[b][1,4]dioxine 95+%
(R)-2-(Bromomethyl)-2,3-dihydrobenzo[b][1,4]dioxine

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12756174
CAS-Nummer: 1142953-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR