1,3-difluoro-2-methyl-4-nitrobenzene 97%

Synonyme: 2,6-Difluoro-3-nitrotoluene,1,3-Difluoro-2-methyl-4-nitrobenzene

BD160926-250MG BD160926-25G BD160926-5G BD160926-10G BD160926-1G
BLDPBD160926-250MGEA 21 EUR
BLDPBD160926-250MG BLDPBD160926-25G BLDPBD160926-5G BLDPBD160926-10G BLDPBD160926-1G
1,3-difluoro-2-methyl-4-nitrobenzene 97%
1,3-difluoro-2-methyl-4-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₅F₂NO₂
Molecular Weight: 173,119 g/mol
Lagertemperatur: Ambient
MDL: MFCD11519030
CAS-Nummer: 79562-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR