4-Bromo-2,6-diethylpyridine 95%

BD160921-1G
BLDPBD160921-1GEA 98 EUR
BLDPBD160921-1G
4-Bromo-2,6-diethylpyridine 95%
4-Bromo-2,6-diethylpyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₂BrN
Molecular Weight: 214,1 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12755732
CAS-Nummer: 877133-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR