N-Acetyl-L-prolinamide 98%

BLDPBD16089-5GEA 37 EUR
BLDPBD16089-5G BLDPBD16089-25G BLDPBD16089-1G BLDPBD16089-250MG
N-Acetyl-L-prolinamide 98%
N-Acetyl-L-prolinamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₁₂N₂O₂
Molecular Weight: 156,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037340
CAS-Nummer: 16395-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR