4-Chloro-6-methylpicolinonitrile 98%

BD160866-1G BD160866-100MG BD160866-250MG
BLDPBD160866-1GEA 259 EUR
BLDPBD160866-1G BLDPBD160866-100MG BLDPBD160866-250MG
4-Chloro-6-methylpicolinonitrile 98%
4-Chloro-6-methylpicolinonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄ClN₃
Molecular Weight: 153,57 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09607719
CAS-Nummer: 104711-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR