tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate 98%

Synonyme: tert-Butyl 2,7-diazaspiro[4.5]decane-2-carboxylate

BD160856-100MG
BLDPBD160856-100MGEA 78 EUR
BLDPBD160856-100MG
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate 98%
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₃H₂₄N₂O₂
Molecular Weight: 240,35 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08234437
CAS-Nummer: 885268-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR