2-(Ethoxycarbonyl)phenylboronsäurepinakolester 97%

BLDPBD16084-25GEA 233 EUR
BLDPBD16084-25G BLDPBD16084-250MG BLDPBD16084-5G BLDPBD16084-1G BLDPBD16084-10G
2-(Ethoxycarbonyl)phenylboronsäurepinakolester 97%
2-(Ethoxycarbonyl)phenylboronsäurepinakolester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03453660
CAS-Nummer: 269409-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR