4-Methyl-β-styrylboronic acid pinacol ester 98%

Synonyme: (E)-4,4,5,5-Tetramethyl-2-(4-methylstyryl)-1,3,2-dioxaborolane

BD16083-5G BD16083-1G BD16083-100MG BD16083-250MG
BLDPBD16083-5GEA 617 EUR
BLDPBD16083-5G BLDPBD16083-1G BLDPBD16083-100MG BLDPBD16083-250MG
4-Methyl-β-styrylboronic acid pinacol ester 98%
4-Methyl-beta-styrylboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₁BO₂
Molecular Weight: 244,14 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD03788749
CAS-Nummer: 149777-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR