5-Acetylpyridin-2(1H)-one 97%

Synonyme: 2-Hydroxy-5-acetylpyridine

BD160821-5G BD160821-10G BD160821-100MG BD160821-1G BD160821-250MG BD160821-25G
BLDPBD160821-5GEA 167 EUR
BLDPBD160821-5G BLDPBD160821-10G BLDPBD160821-100MG BLDPBD160821-1G BLDPBD160821-250MG BLDPBD160821-25G
5-Acetylpyridin-2(1H)-one 97%
5-Acetylpyridin-2(1H)-one

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇NO₂
Molecular Weight: 137,1400 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10697705
CAS-Nummer: 1124-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR