2,3-Difluoro-4-n-propoxybenzeneboronic acid 98%

BLDPBD160705-1GEA 17 EUR
BLDPBD160705-1G BLDPBD160705-5G BLDPBD160705-25G
2,3-Difluoro-4-n-propoxybenzeneboronic acid 98%
2,3-Difluoro-4-n-propoxybenzeneboronic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₁BF₂O₃
Molecular Weight: 215,991 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14584457
CAS-Nummer: 212837-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR