4'-Propyl-4-biphenylboronic acid 98%

BLDPBD160702-1GEA 13 EUR
BLDPBD160702-1G BLDPBD160702-5G BLDPBD160702-25G
4'-Propyl-4-biphenylboronic acid 98%
4'-Propyl-4-biphenylboronic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₇BO₂
Molecular Weight: 240,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01114670
CAS-Nummer: 153035-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR