Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid 95%

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-iodophenyl)butanoic acid

BD16066-1G BD16066-100MG BD16066-5G BD16066-250MG
BLDPBD16066-1GEA 153 EUR
BLDPBD16066-1G BLDPBD16066-100MG BLDPBD16066-5G BLDPBD16066-250MG
Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid 95%
Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₅H₂₀INO₄
Molecular Weight: 405,24 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01860963
CAS-Nummer: 269396-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR