4-Hydroxy-3-methylphenylacetic acid 95%

BLDPBD160659-100MGEA 63 EUR
BLDPBD160659-100MG BLDPBD160659-5G BLDPBD160659-1G BLDPBD160659-250MG
4-Hydroxy-3-methylphenylacetic acid 95%
4-Hydroxy-3-methylphenylacetic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06656118
CAS-Nummer: 29121-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR