3,5-Dibromo-4-fluoroaniline 96%

BD160635-100MG BD160635-250MG BD160635-1G BD160635-5G
BLDPBD160635-100MGEA 39 EUR
BLDPBD160635-100MG BLDPBD160635-250MG BLDPBD160635-1G BLDPBD160635-5G
3,5-Dibromo-4-fluoroaniline 96%
3,5-Dibromo-4-fluoroaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₄Br₂FN
Molecular Weight: 268,91 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09835195
CAS-Nummer: 1003709-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR