Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%

Synonyme: 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride,1-Benzyl-3-ethoxycarbonyl-4-piperidone Hydrochloride,1-Benzyl-4-oxo-3-piperidinecarboxylic acid ethyl ester hydrochloride

BD16055-25G BD16055-100G BD16055-500G BD16055-5G BD16055-10G
BLDPBD16055-25GEA 16 EUR
BLDPBD16055-25G BLDPBD16055-100G BLDPBD16055-500G BLDPBD16055-5G BLDPBD16055-10G
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₅H₁₉NO₃·HCl
Molecular Weight: 297,780 g/mol
Lagertemperatur: Ambient
MDL: MFCD00012800
CAS-Nummer: 1454-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR