2-Morpholinopyrimidin-5-ylboronic acid 95%

Synonyme: 2-(Morpholine-4-yl)pyrimidine-5-boronic acid

BD160526-5G BD160526-1G BD160526-250MG
BLDPBD160526-5GEA 276 EUR
BLDPBD160526-5G BLDPBD160526-1G BLDPBD160526-250MG
2-Morpholinopyrimidin-5-ylboronic acid 95%
2-Morpholinopyrimidin-5-ylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₁₂BN₃O₃
Molecular Weight: 209,01 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07375148
CAS-Nummer: 870521-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR