Methyl 3-amino-2,6-difluorobenzoate 98%

Synonyme: 2,4-Difluoro-3-(methoxycarbonyl)aniline

BD160525-5G BD160525-25G BD160525-1G
BLDPBD160525-5GEA 56 EUR
BLDPBD160525-5G BLDPBD160525-25G BLDPBD160525-1G
Methyl 3-amino-2,6-difluorobenzoate 98%
Methyl 3-amino-2,6-difluorobenzoate

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₇F₂NO₂
Molecular Weight: 187,143 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD13195461
CAS-Nummer: 84832-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR