2-(Bromomethyl)-4-phenylthiazole 95%

BD160521-100MG BD160521-1G BD160521-250MG
BLDPBD160521-100MGEA 74 EUR
BLDPBD160521-100MG BLDPBD160521-1G BLDPBD160521-250MG
2-(Bromomethyl)-4-phenylthiazole 95%
2-(Bromomethyl)-4-phenylthiazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈BrNS
Molecular Weight: 254,15 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12405586
CAS-Nummer: 78502-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR