(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid 97%

BD16052-1G BD16052-250MG BD16052-5G BD16052-10G
BLDPBD16052-1GEA 51 EUR
BLDPBD16052-1G BLDPBD16052-250MG BLDPBD16052-5G BLDPBD16052-10G
(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid 97%
(S)-2-(((Benzyloxy)carbonyl)amino)-5-ethoxy-5-oxopentanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₉NO₆
Molecular Weight: 309,31 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00274757
CAS-Nummer: 35726-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR