Methyl-2-methoxy-5-nitrobenzoat 95%

Synonyme: Methyl 2-methoxy-5-nitrobenzoate

BD160487-5G BD160487-100G BD160487-25G BD160487-1G
BLDPBD160487-5GEA 41 EUR
BLDPBD160487-5G BLDPBD160487-100G BLDPBD160487-25G BLDPBD160487-1G
Methyl-2-methoxy-5-nitrobenzoat 95%
Methyl-2-methoxy-5-nitrobenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉NO₅
Molecular Weight: 211,18 g/mol
Lagertemperatur: Ambient
MDL: MFCD00673023
CAS-Nummer: 34841-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR