3-Amino-5-cyclobutyl-1H-pyrazole 97%

BLDPBD160478-100MGEA 23 EUR
BLDPBD160478-100MG BLDPBD160478-1G BLDPBD160478-250MG BLDPBD160478-5G
3-Amino-5-cyclobutyl-1H-pyrazole 97%
3-Amino-5-cyclobutyl-1H-pyrazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₁₁N₃
Lagertemperatur: Kühlschrank
MDL: MFCD08061045
CAS-Nummer: 326827-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR