2-Brom-3-iodpyridin 98%

BD160476-250MG BD160476-100MG BD160476-5G BD160476-25G BD160476-1G BD160476-100G BD160476-10G
BLDPBD160476-250MGEA 8 EUR
BLDPBD160476-250MG BLDPBD160476-100MG BLDPBD160476-5G BLDPBD160476-25G BLDPBD160476-1G BLDPBD160476-100G BLDPBD160476-10G
2-Brom-3-iodpyridin 98%
2-Bromo-3-iodopyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
Lagertemperatur: Ambient
MDL: MFCD09056782
CAS-Nummer: 265981-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR