7-Methoxyquinolin-3-amine 97%

BD160429-1G BD160429-250MG BD160429-100MG
BLDPBD160429-1GEA 442 EUR
BLDPBD160429-1G BLDPBD160429-250MG BLDPBD160429-100MG
7-Methoxyquinolin-3-amine 97%
7-Methoxyquinolin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,20 g/mol
Lagertemperatur: Ambient
MDL: MFCD01246355
CAS-Nummer: 87199-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR