6-Chloroquinolin-3-amine 98%

BD160426-250MG BD160426-100MG
BLDPBD160426-250MGEA 256 EUR
BLDPBD160426-250MG BLDPBD160426-100MG
6-Chloroquinolin-3-amine 98%
6-Chloroquinolin-3-amine

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₇ClN₂
Molecular Weight: 178,61 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11917897
CAS-Nummer: 1051372-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR