8-Bromoisoquinolin-1(2H)-one 95%

BLDPBD160371-1GEA 128 EUR
BLDPBD160371-1G BLDPBD160371-250MG BLDPBD160371-100MG BLDPBD160371-10G BLDPBD160371-25G BLDPBD160371-5G
8-Bromoisoquinolin-1(2H)-one 95%
8-Bromoisoquinolin-1(2H)-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858429
CAS-Nummer: 475994-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR