5-Bromo-2-chloro-4-methylpyrimidine 97%

BD160370-25G BD160370-500G BD160370-250MG BD160370-5G BD160370-10G BD160370-1G BD160370-100G
BLDPBD160370-25GEA 229 EUR
BLDPBD160370-25G BLDPBD160370-500G BLDPBD160370-250MG BLDPBD160370-5G BLDPBD160370-10G BLDPBD160370-1G BLDPBD160370-100G
5-Bromo-2-chloro-4-methylpyrimidine 97%
5-Bromo-2-chloro-4-methylpyrimidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₄BrClN₂
Molecular Weight: 207,4557 g/mol
Lagertemperatur: Ambient
MDL: MFCD13193654
CAS-Nummer: 633328-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR