4-(Trifluoromethyl)pyrimidine-5-carboxylic acid 95%

BD160347-1G BD160347-250MG BD160347-100MG BD160347-10G BD160347-5G
BLDPBD160347-1GEA 112 EUR
BLDPBD160347-1G BLDPBD160347-250MG BLDPBD160347-100MG BLDPBD160347-10G BLDPBD160347-5G
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid 95%
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₃F₃N₂O₂
Molecular Weight: 192,095 g/mol
Lagertemperatur: Ambient
MDL: MFCD11106713
CAS-Nummer: 220880-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR