Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid 95%

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-dichlorophenyl)butanoic acid

BD16034-250MG BD16034-100MG
BLDPBD16034-250MGEA 73 EUR
BLDPBD16034-250MG BLDPBD16034-100MG
Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid 95%
Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₉Cl₂NO₄
Molecular Weight: 348,23 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01860946
CAS-Nummer: 269396-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR