5-Bromo-3-methyl-1H-pyrazolo[3,4-c]pyridine 97%

BD160330-250MG BD160330-100MG BD160330-5G BD160330-1G
BLDPBD160330-250MGEA 68 EUR
BLDPBD160330-250MG BLDPBD160330-100MG BLDPBD160330-5G BLDPBD160330-1G
5-Bromo-3-methyl-1H-pyrazolo[3,4-c]pyridine 97%
5-Bromo-3-methyl-1H-pyrazolo[3,4-c]pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BrN₃
Molecular Weight: 212,0500 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12024520
CAS-Nummer: 929617-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR