4-Bromo-2-cyanobenzeneacetonitrile 95%

BD160312-250MG BD160312-1G BD160312-5G
BLDPBD160312-250MGEA 56 EUR
BLDPBD160312-250MG BLDPBD160312-1G BLDPBD160312-5G
4-Bromo-2-cyanobenzeneacetonitrile 95%
4-Bromo-2-cyanobenzeneacetonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅BrN₂
Molecular Weight: 221,0500 g/mol
Lagertemperatur: Ambient
MDL: MFCD09999218
CAS-Nummer: 925672-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR