7-Nitro-2(1H)-quinoxalinone 97%

BD160311-5G BD160311-250MG BD160311-100MG BD160311-1G BD160311-25G
BLDPBD160311-5GEA 363 EUR
BLDPBD160311-5G BLDPBD160311-250MG BLDPBD160311-100MG BLDPBD160311-1G BLDPBD160311-25G
7-Nitro-2(1H)-quinoxalinone 97%
7-Nitro-2(1H)-quinoxalinone

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅N₃O₃
Molecular Weight: 191,1436 g/mol
Lagertemperatur: Ambient
MDL: MFCD09999191
CAS-Nummer: 89898-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR