5-Bromo-2-ethylbenzoic acid 98%

BD160280-1G BD160280-250MG BD160280-100MG
BLDPBD160280-1GEA 275 EUR
BLDPBD160280-1G BLDPBD160280-250MG BLDPBD160280-100MG
5-Bromo-2-ethylbenzoic acid 98%
5-Bromo-2-ethylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
Lagertemperatur: Ambient
MDL: MFCD16619423
CAS-Nummer: 439937-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR